Chloroform-[D₁] NMR, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
MQ200
151831-100G 151831-250G 151831-50G
SIAL151831-100GEA 63.3 EUR
SIAL151831-100G SIAL151831-250G SIAL151831-50G
Chloroform-[D₁] NMR, Sigma-Aldrich®
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1 III