Methanol-[D₄], Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
M-Clarity™ quality level = MQ200
343803-10G
343803-50G
343803-5G
SIAL343803-10GEA
188
EUR
SIAL343803-10G
SIAL343803-50G
SIAL343803-5G
Methanol-[D₄], Sigma-Aldrich®
Methanol-[D4]
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 Melting Pt: –99 °C Density: 0.888 g/cm³ (20 °C) Flash Pt: 11 Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3 II |