Pyridine-[D₅], Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Pyridine-[²H₅]
M-Clarity™ quality level = MQ200
532967-25G 532967-5G
SIAL532967-25GEA 635 EUR
SIAL532967-25G SIAL532967-5G
Pyridine-[D₅], Sigma-Aldrich®
Pyridine-[D5]
Formula: C₅D₅N
MW: 84.06 g/mol
Boiling Pt: 114.4
Melting Pt: –41 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 20
Storage Temperature: Ambient
MDL Number: MFCD00044639
CAS Number: 7291-22-7
EINECS: 230-720-2
UN: 1282
ADR: 3 II