Acetone-[D₆], Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL175862-5GEA
180
EUR
SIAL175862-5G
Acetone-[D₆], Sigma-Aldrich®
Acetone-[D₆]
Formula:
D₃CC(O)CD₃ MW: 64,03 g/mol Boiling Pt: 55,5 °C (1013 hPa) Melting Pt: –95,4 °C Density: 0,79 g/cm³ (20 °C) Flash Pt: –19 °C (closed cup) |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 UN: 1090 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...