N,N-Dimethylformamide-[D₇] 99.5 atom % D NMR
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7
183600010 183600100
ACRO183600010EA 81 EUR
ACRO183600010 ACRO183600100
N,N-Dimethylformamide-[D₇] 99.5 atom % D NMR
N,N-Dimethylformamide-[D7]
Formula: DCOON(CD₃)₂
MW: 80.04 g/mol
Boiling Pt: 152
Melting Pt: –60 °C
Density: 1.03 g/cm³
Flash Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
GC ≥99.0 %
D-Enrichment ≥99.4 %