Iodomethane[D₃] (99.5% D) stabilised, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl iodide-D3, Iodomethane-[²H₃]
Stabilization: Stabilised with copper
M-Clarity™ quality level = MQ100
176036-1G 176036-25G 176036-50G 176036-5G
SIAL176036-1GEA 50.2 EUR
SIAL176036-1G SIAL176036-25G SIAL176036-50G SIAL176036-5G
Iodomethane[D₃] (99.5% D) stabilised, Sigma-Aldrich®
Iodomethane-[D3]
Formula: D₃CI
Boiling Pt: 42 °C (1013 hPa)
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00001074
CAS Number: 865-50-9
EINECS: 212-744-5
UN: 2644
ADR: 6.1,I