Methanol-[D₃], Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Carbinol-D3, Hydroxy methane-D3, Methanol-[²H₃], Wood alcohol-D3, Methyl alcohol-D3
M-Clarity™ quality level = MQ200
343854-10G 343854-1G 343854-5G
SIAL343854-10GEA 418 EUR
SIAL343854-10G SIAL343854-1G SIAL343854-5G
Methanol-[D₃], Sigma-Aldrich®
Methanol-[D3]
Formula: D₃COH
MW: 35.02 g/mol
Boiling Pt: 64.5
Melting Pt: –98 °C
Density: 0.867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00055877
CAS Number: 1849-29-2
EINECS: 217-435-9
UN: 1230
ADR: 3 II