1,2-Dichlorobenzene-[D₄], Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Dichlorobenzene-[²H₄]
M-Clarity™ quality level = MQ100
331511-10G 331511-1G 331511-5G
SIAL331511-10GEA 482 EUR
SIAL331511-10G SIAL331511-1G SIAL331511-5G
1,2-Dichlorobenzene-[D₄], Sigma-Aldrich®
1,2-Dichlorobenzene-[D4]
Formula: C₆D₄Cl₂
MW: 150.97 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037106
CAS Number: 2199-69-1
EINECS: 218-606-0
UN: 1591
ADR: 6.1,III