Chlorobenzene-[D₅], Sigma-Aldrich®

Supplier: Merck

Synonyms: Chlorobenzene-[²H₅], Phenylchloride-D5

M-Clarity™ quality level = MQ200

176605-1G 176605-5G
SIAL176605-1GEA 128 EUR
SIAL176605-1G SIAL176605-5G
Chlorobenzene-[D₅], Sigma-Aldrich®
Chlorobenzene-[D5]
Formula: C₆D₅Cl
MW: 117.52 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –46 °C
Density: 1.15 g/cm³ (20 °C)
MDL Number: MFCD00000531
CAS Number: 3114-55-4
EINECS: 221-482-0
UN: 1134
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR