Cyclohexanol-[D12], Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Hydroxycyclohexane-D12
M-Clarity™ quality level = MQ200
175773-1G 175773-5G
SIAL175773-1GEA 141 EUR
SIAL175773-1G SIAL175773-5G
Cyclohexanol-[D12], Sigma-Aldrich®
Cyclohexanol-[D12]
Formula: C₆D₁₂O
Boiling Pt: 157.4 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.07 g/cm³ (20 °C)
CAS Number: 66522-78-9
EINECS: 266-389-6