Toluene-[D₈], Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylmethane-D8, Methylbenzene-D8, Toluene-[²H₈]
MQ100
434388-10G 434388-25G 434388-5G
SIAL434388-10GEA 137 EUR
SIAL434388-10G SIAL434388-25G SIAL434388-5G
Toluene-[D₈], Sigma-Aldrich®
Toluene-[D8]
Formula: C₆D₅CD₃
MW: 100.08 g/mol
Boiling Pt: 110
Melting Pt: –85 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 4 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044638
CAS Number: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3 II