Deuterium oxide

Supplier: Roth Carl

Synonyms: Water-D2, Heavy water

6672.1 6672.2 6672.3 6672.4
ROTH6672.1EA 0 EUR
ROTH6672.1 ROTH6672.2 ROTH6672.3 ROTH6672.4
Deuterium oxide
Deuterium oxide
Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR