Deuterium oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL293040-100GEA 265 EUR
SIAL293040-100G SIAL293040-25G
Deuterium oxide, Sigma-Aldrich®
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101,3 °C (1013 hPa)
Melting Pt: 3,8 °C
Density: 1,1 g/cm³ (20 °C)
MDL Number: MFCD00044636
CAS Number: 7789-20-0
Merck Index: 12,02984

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR