Fullerene C60, Sigma-Aldrich®
Supplier: Merck
Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
M-Clarity™ quality level = MQ100
572500-1G 572500-250MG 572500-5G
SIAL572500-1GEA 621 EUR
SIAL572500-1G SIAL572500-250MG SIAL572500-5G
Fullerene C60, Sigma-Aldrich®
Fullerene C60
Formula: C₆₀
MW: 720.66 g/mol
Melting Pt: > 280 °C
Density: ∼1.65 g/cm³ (20 °C)
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM