Fullerene C70, Sigma-Aldrich®
Supplier: Merck
Synonyms: Buckminster-Fullerene C70, [5,6]-Fullerene-C70
M-Clarity™ quality level = MQ100
482994-100MG 482994-500MG
SIAL482994-100MGEA 187 EUR
SIAL482994-100MG SIAL482994-500MG
Fullerene C70, Sigma-Aldrich®
Fullerene C70
Formula: C₇₀
MW: 840.77 g/mol
Melting Pt: > 280 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00282904
CAS Number: 115383-22-7
UN: 1325
ADR: 4.1,III