Benzalkonium chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
12060-100G 12060-500G 12060-1KG 12060-5G
SIAL12060-100GEA 46.5 EUR
SIAL12060-100G SIAL12060-500G SIAL12060-1KG SIAL12060-5G
Benzalkonium chloride, Sigma-Aldrich®
Benzalkonium chloride
Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Melting Pt: 29…34 °C
Density: 0.98 g/cm³ (20 °C)
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8,III