Boron trifluoride diethyl ether complex, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
M-Clarity™ quality level = MQ200
216607-100ML 216607-800ML
SIAL216607-100MLEA 90.8 EUR
SIAL216607-100ML SIAL216607-800ML
Boron trifluoride diethyl ether complex, Sigma-Aldrich®
Boron trifluoride diethyl ether complex
Formula: C₄H₁₀BF₃O
MW: 141.93 g/mol
Boiling Pt: 126
Melting Pt: –60 °C
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 59
Storage Temperature: Ambient
MDL Number: MFCD00013194
CAS Number: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8 I
Merck Index: 13,01340