Tungsten hexacarbonyl, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Tungsten carbonyl, Hexacarbonyltungsten
M-Clarity™ quality level = MQ100
472956-25G
472956-5G
SIAL472956-25GEA
825
EUR
SIAL472956-25G
SIAL472956-5G
Tungsten hexacarbonyl, Sigma-Aldrich®
Tungsten hexacarbonyl
Formula:
C₆O₆W MW: 351.9 g/mol Boiling Pt: 451.3 Melting Pt: 169…171 °C Density: 2.65 g/cm³ (20 °C) |
MDL Number:
MFCD00011462 CAS Number: 14040-11-0 EINECS: 237-880-2 UN: 3466 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...