L(+)-Lactic acid 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
69780-1L
69780-250ML
SIAL69780-1LEA
166
EUR
SIAL69780-1L
SIAL69780-250ML
L(+)-Lactic acid 40% in aqueous solution, Sigma-Aldrich®
L(+)-Lactic acid
Formula:
C₃H₆O₃ MW: 90.08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00064266 CAS Number: 79-33-4 EINECS: 201-196-2 |