D-(+)-Glucose monohydrate, powder, TECHNICAL
Supplier: VWR Chemicals
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
24374.366EA 125 EUR
24374.366 24374.460
D-(+)-Glucose monohydrate, powder, TECHNICAL
D(+)-Glucose monohydrate
Formula: C₆H₁₂O₆·H₂O
Melting Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149450
CAS Number: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Water 7.0 - 9.5 %