D-(+)-Glucose monohydrate, Millipore®
Supplier: Merck
Synonyms:
(2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
MQ200
49159-1KG
49159-5KG
SIAL49159-1KGEA
42.6
EUR
SIAL49159-1KG
SIAL49159-5KG
D-(+)-Glucose monohydrate, Millipore®
D(+)-Glucose monohydrate
Formula:
C₆H₁₂O₆·H₂O MW: 198.17 g/mol Melting Pt: 83 Storage Temperature: Ambient |
MDL Number:
MFCD00149450 CAS Number: 14431-43-7 EINECS: 200-075-1 |