D-(+)-Glucose monohydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALD9559-100GEA 70.7 EUR
SIALD9559-100G SIALD9559-1KG
D-(+)-Glucose monohydrate, Sigma-Aldrich®
D-(+)-Glucose monohydrate
Formula: C₆H₁₂O₆·H₂O
MW: 198,17 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00149450
CAS Number: 14431-43-7
https://si.vwr.com/stibo/web/std.lang.all/21/23/2302123.jpg