Blood collection needles, S-Monovette®
Supplier: DESAGA
14.1205 85.1160 85.1162
DESG14.1205EA 337 EUR
DESG14.1205 DESG85.1160 DESG85.1162
Blood collection needles, S-Monovette®
Syringe Needles
These S-Monovette®-Needles are suitable for the S-Monovette® blood collection system.
  • Membrane screw cap connection
  • Sterile and pyrogen-free
Certifikace: CE0124 certified.
Balení: Individual sterile packaging.