D-(+)-Sucrose, Sigma-Aldrich®

Supplier: Merck

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

M-Clarity™ quality level = MQ200

16104-2.5KG 16104-1KG 16104-250G 16104-25KG 16104-6X250G
SIAL16104-2.5KGEA 110 EUR
SIAL16104-2.5KG SIAL16104-1KG SIAL16104-250G SIAL16104-25KG SIAL16104-6X250G
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR