D-(+)-Galactose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
G5388-100G G5388-1KG G5388-500G
SIALG5388-100GEA 92 EUR
SIALG5388-100G SIALG5388-1KG SIALG5388-500G
D-(+)-Galactose, Sigma-Aldrich®
D(+)-Galactose
Formula: C₆H₁₂O₆
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 163…169 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM