D-(+)-Mannose, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
MQ200
M2069-100G M2069-25G M2069-500G
SIALM2069-100GEA 192 EUR
SIALM2069-100G SIALM2069-25G SIALM2069-500G
D-(+)-Mannose, Sigma-Aldrich®
D(+)-Mannose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 133
Density: 1.539 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772