D-(+)-Xylose, Sigma-Aldrich®
Supplier: Merck
95729-100G 95729-25G 95729-500G
SIAL95729-100GEA 164 EUR
SIAL95729-100G SIAL95729-25G SIAL95729-500G
D-(+)-Xylose, Sigma-Aldrich®
D(+)-Xylose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 154
Density: 1.525 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7
Merck Index: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM