D-(-)-Fructose, Sigma-Aldrich®
Supplier: Merck
Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
M-Clarity™ quality level = MQ200
F3510-100G F3510-500G F3510-5KG
SIALF3510-100GEA 43 EUR
SIALF3510-100G SIALF3510-500G SIALF3510-5KG
D-(-)-Fructose, Sigma-Aldrich®
D(-)-Fructose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 552
Melting Pt: 100…110 °C
Density: 1.59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM