Cellulose, Sigma-Aldrich®
Supplier: Merck
Synonyms: Microcrystallinecellulose
M-Clarity™ quality level = MQ100
310697-1KG 310697-500G 310697-50G
SIAL310697-1KGEA 128 EUR
SIAL310697-1KG SIAL310697-500G SIAL310697-50G
Cellulose, Sigma-Aldrich®
Cellulose
Formula: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
Melting Pt: 260…270 °C
Density: 1.27…1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977