Agarose for electrophoresis low electroendosmosis (EEO) (Mr <= 0.10)
Supplier: Thermo Fisher Scientific
159820100 159820250 159821000
ACRO159820100EA 34.8 EUR
ACRO159820100 ACRO159820250 ACRO159821000
Agarose for electrophoresis low electroendosmosis (EEO) (Mr <= 0.10)
Agarose
Melting Pt: 60 - 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Gel strength ≥800 g/cm² (1% gel)
Gel point 34 to 45°C
Electroendosmosis ≤0.12 (-Mr)
Sulfate (SO4) ≤0.3 %