Agarose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A4718-100G A4718-25G
SIALA4718-100GEA 550 EUR
SIALA4718-100G SIALA4718-25G
Agarose, Sigma-Aldrich®
Agarose
Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM