Starch, soluble, Sigma-Aldrich®
Supplier: Merck
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
M-Clarity™ quality level = MQ100
33615-1KG 33615-250G
SIAL33615-1KGEA 267 EUR
SIAL33615-1KG SIAL33615-250G
Starch, soluble, Sigma-Aldrich®
Starch, soluble
Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878