Starch, soluble, Sigma-Aldrich®
Supplier: Merck
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
MQ200
S9765-100G S9765-1KG S9765-250G S9765-500G
SIALS9765-100GEA 36.2 EUR
SIALS9765-100G SIALS9765-1KG SIALS9765-250G SIALS9765-500G
Starch, soluble, Sigma-Aldrich®
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878