Lactic acid 10% in aqueous solution
Supplier: THERMO FISHER DIAGNOSTICS
Danger
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
SR0021H SR0021K
OXOISR0021HEA 60.5 EUR
OXOISR0021H OXOISR0021K
Lactic acid 10% in aqueous solution
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0