Lactic acid 85% 85 aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
MQ100
L1250-1L L1250-500ML
SIALL1250-1LEA 98 EUR
SIALL1250-1L SIALL1250-500ML
Lactic acid 85% 85 aqueous solution, Sigma-Aldrich®
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Merck Index: 13,05351