Lactic acid ≥85% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W261106-10KG-K W261106-1KG-K W261106-25KG-K
SIALW261106-10KG-KEA 349 EUR
SIALW261106-10KG-K SIALW261106-1KG-K SIALW261106-25KG-K
Lactic acid ≥85% in aqueous solution, Sigma-Aldrich®
Lactic acid
Formula: H₃CCH(OH)COOH
Boiling Pt: 122 °C (20 hPa)
Density: 1.11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0