DL-Sodium lactate 60% w/w aqueous solution
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate
41529.AK 41529.K2
41529.AKEA 72.6 EUR
41529.AK 41529.K2
DL-Sodium lactate 60% w/w aqueous solution
DL-Sodium lactate
CAS No.: 72-17-3 Molecular Formula: CH3CH(OH)CO2Na Formula Weight: 112.06 Physical Form: Liquid Density: 1.326 Appearance: Colourless Odour: Odourless Beilstein Reference: 4332999 Merck Reference: 14,8635 MDL No.: MFCD00065400
Useful chiral synthon; building block for depsipeptides
Formula: H₃CCH(OH)COONa
MW: 112.06 g/mol
Density: 1.326
Storage Temperature: Ambient
MDL Number: MFCD00065400
CAS Number: 72-17-3
EINECS: 200-772-0
Merck Index: 14,8635