Acrylamide : Bisacrylamide (29:1) 40% aqueous solution
Supplier: PanReac AppliChem
Danger
Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis
A0385.0500 A0385.1000
A0385.0500EA 71.6 EUR
A0385.0500 A0385.1000
Acrylamide : Bisacrylamide (29:1) 40% aqueous solution
Acrylamide-Bis (32:1)
Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1 III