Nickel chromium alloy (80:20 w%), wire, Ø 0.05 mm (0.002 in)
Supplier: Thermo Scientific
|
Warning
|
44187.H9
44187.J5
44187.H9EA
277
EUR
44187.H9
44187.J5
Nickel chromium alloy (80:20 w%), wire, Ø 0.05 mm (0.002 in)
Nickel Chromium alloy (80:20 w%)
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈0.017g/m
Odour: Odourless
MDL No.: MFCD00673110
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS Number: 11106-97-1 |