Gelatin
Supplier: Roth Carl
4274.1 4274.2
ROTH4274.1EA 56.8 EUR
ROTH4274.1 ROTH4274.2
Gelatin
Proteins and Peptides
Order Now
SPECIFICATIONS
Protein/peptide name Gelatin
Protein synonyms Gelatine
CAS No. 9000-70-8
Storage conditions Ambient