Polydimethylsiloxane, 1000 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ200
378399-1L 378399-250ML
SIAL378399-1LEA 376 EUR
SIAL378399-1L SIAL378399-250ML
Polydimethylsiloxane, 1000 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Clear colourless low-volatility polydimethylsiloxane
Delivery information: This product is not available in all countries. Please check with your local VWR International office or supplier.
Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Boiling Pt: >140 °C (0.002 mmHg)) (lit.)
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 321 °C (closed cup)
MDL Number: MFCD00084411
CAS Number: 63148-62-9