Polydimethylsiloxane, 5 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ100
317667-5ML 317667-1L 317667-250ML
SIAL317667-5MLEA 65.8 EUR
SIAL317667-5ML SIAL317667-1L SIAL317667-250ML
Polydimethylsiloxane, 5 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Clear colourless hexamethyldisiloxane
Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Density: 0.98 g/cm³ (25 °C)
Flash Pt: 321 °C (closed cup)
MDL Number: MFCD00084411
CAS Number: 63148-62-9