Polyethylene glycol 1,000
Supplier: Thermo Scientific
Synonyms: PEG 1,000, PEG 900-1100
192250010 192250051
ACRO192250010EA 52.3 EUR
ACRO192250010 ACRO192250051
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Formula: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 35…40 °C
Density: 1,2 g/cm³ (20 °C)
Flash Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD01779605
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 13,07651
Order Now
Specification Test Results
Appearance White Waxy solid
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Hydroxyl value 107 to 118 mg KOH/g