Polyethylene glycol 1,000, Sigma-Aldrich®
Supplier: Merck
Synonyms:
PEG 1,000, PEG 900-1100
M-Clarity™ quality level = MQ200
81188-1KG
81188-250G
SIAL81188-1KGEA
140
EUR
SIAL81188-1KG
SIAL81188-250G
Polyethylene glycol 1,000, Sigma-Aldrich®
Polyethylene glycol 1,000
Formula:
H(OCH₂CH₂)nOH MW: 1000 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 33…40 °C Density: 1.2 g/cm³ (20 °C) Flash Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 13,07651 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...