Polyethylene glycol 1,000, Sigma-Aldrich®

Supplier: Merck

Synonyms: PEG 1,000, PEG 900-1100

M-Clarity™ quality level = MQ200

81189-100G 81189-1KG 81189-250G
SIAL81189-100GEA 45.4 EUR
SIAL81189-100G SIAL81189-1KG SIAL81189-250G
Polyethylene glycol 1,000, Sigma-Aldrich®
Polyethylene glycol 1,000
Formula: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 33…40 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 13,07651

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR