Polyethylene glycol 300, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 300
90878-1L-F 90878-250ML-F
SIAL90878-1L-FEA 89.2 EUR
SIAL90878-1L-F SIAL90878-250ML-F
Polyethylene glycol 300, Sigma-Aldrich®
Polyethylene glycol 300
Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Melting Pt: –15…–10 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2