Polyethylene glycol 4,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 4,000
95904-1KG-F 95904-250G-F
SIAL95904-1KG-FEA 112 EUR
SIAL95904-1KG-F SIAL95904-250G-F
Polyethylene glycol 4,000, Sigma-Aldrich®
Polyethylene glycol 4,000
Formula: H(OCH₂CH₂)nOH
MW: 4000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 53…58 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779612
CAS Number: 25322-68-3
EINECS: 500-038-2