Polyethylene glycol 6,000
Supplier: Thermo Fisher Scientific
A17541.0B
A17541.0I
A17541.30
A17541.0BEA
76.2
EUR
A17541.0B
A17541.0I
A17541.30
Polyethylene glycol 6,000
Polyethylene glycol 6,000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Odour: Odourless
pH: 5-7
Melting Point: 60-63°
Merck Reference: 14,7568
MDL No.: MFCD01779614
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 60…63 °C Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01779614 CAS Number: 25322-68-3 EINECS: 203-473-3 Merck Index: 14,7568 |