Polyethylene glycol 6,000, TECHNICAL

Supplier: VWR Chemicals

Synonyms: PEG 6,000, PEG 6000

26603.293EA 53.4 EUR
26603.293 26603.460
Polyethylene glycol 6,000, TECHNICAL
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Boiling Pt: >250 °C
Melting Pt: 60…63 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Hydroxyl value 16 - 22
Solidification point 55 - 61 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR