Polyethylene glycol 6,000, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
81253-1KG 81253-2.5KG 81253-250G
SIAL81253-1KGEA 228 EUR
SIAL81253-1KG SIAL81253-2.5KG SIAL81253-250G
Polyethylene glycol 6,000, Sigma-Aldrich®
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 55…62 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2